Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614773 MIM: 114106 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSS51 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MSS51 - MSS51 mitochondrial translational activator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP3CB - protein phosphatase 3 catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142353.2 | 2659 | UTR 3 | NP_001135825.1 | |||
NM_001142354.2 | 2659 | UTR 3 | NP_001135826.1 | |||
NM_001289968.1 | 2659 | Intron | NP_001276897.1 | |||
NM_001289969.1 | 2659 | Intron | NP_001276898.1 | |||
NM_021132.3 | 2659 | UTR 3 | NP_066955.1 | |||
XM_005269944.1 | 2659 | UTR 3 | XP_005270001.1 | |||
XM_017016379.1 | 2659 | UTR 3 | XP_016871868.1 |
Set Membership: |
HapMap Validated |