Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164785 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MDM2 - MDM2 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145337.2 | Intron | NP_001138809.1 | ||||
NM_001145339.2 | Intron | NP_001138811.1 | ||||
NM_001145340.2 | Intron | NP_001138812.1 | ||||
NM_001278462.1 | Intron | NP_001265391.1 | ||||
NM_002392.5 | Intron | NP_002383.2 | ||||
XM_005268872.4 | Intron | XP_005268929.1 | ||||
XM_006719399.3 | Intron | XP_006719462.1 | ||||
XM_006719400.3 | Intron | XP_006719463.1 |
Set Membership: |
HapMap |