Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGCTCTTTTGGATTGGATACT[A/G]TCTGTGATCACAACTGATACTTGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164785 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130075 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100130075 - SUZ RNA binding domain containing 1 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDM2 - MDM2 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145337.2 | Intron | NP_001138809.1 | ||||
NM_001145339.2 | Intron | NP_001138811.1 | ||||
NM_001145340.2 | Intron | NP_001138812.1 | ||||
NM_001278462.1 | Intron | NP_001265391.1 | ||||
NM_002392.5 | Intron | NP_002383.2 | ||||
XM_005268872.4 | Intron | XP_005268929.1 | ||||
XM_006719399.3 | Intron | XP_006719462.1 | ||||
XM_006719400.3 | Intron | XP_006719463.1 |
Set Membership: |
HapMap Validated |