Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611627 MIM: 606969 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBIL5P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DBIL5P - diazepam binding inhibitor-like 5, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM57A - family with sequence similarity 57 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GEMIN4 - gem nuclear organelle associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015721.2 | 3463 | Silent Mutation | NP_056536.2 | |||
XM_005256667.4 | 3463 | Silent Mutation | XP_005256724.1 | |||
XM_005256670.4 | 3463 | Silent Mutation | XP_005256727.1 | |||
XM_011523910.2 | 3463 | Silent Mutation | XP_011522212.1 | |||
XM_011523911.2 | 3463 | Silent Mutation | XP_011522213.1 | |||
XM_011523912.2 | 3463 | Silent Mutation | XP_011522214.1 | |||
XM_011523913.2 | 3463 | Silent Mutation | XP_011522215.1 | |||
XM_017024709.1 | 3463 | Silent Mutation | XP_016880198.1 |