Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600466 MIM: 602141 MIM: 604592 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ALDH3B1 - aldehyde dehydrogenase 3 family member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4691 - microRNA 4691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7113 - microRNA 7113 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS8 - NADH:ubiquinone oxidoreductase core subunit S8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002496.3 | Intron | NP_002487.1 | ||||
XM_005274013.1 | Intron | XP_005274070.1 | ||||
XM_005274014.2 | Intron | XP_005274071.1 | ||||
XM_011545053.2 | Intron | XP_011543355.1 |
TCIRG1 - T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |