Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607764 MIM: 611052 MIM: 601485 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD3B7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD3B7 - hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142777.1 | 4442 | Intron | NP_001136249.1 | |||
NM_001142778.1 | 4442 | Intron | NP_001136250.1 | |||
NM_025193.3 | 4442 | Intron | NP_079469.2 | |||
XM_005255601.3 | 4442 | Intron | XP_005255658.2 | |||
XM_011545960.2 | 4442 | Intron | XP_011544262.1 | |||
XM_011545961.1 | 4442 | Intron | XP_011544263.1 | |||
XM_011545962.2 | 4442 | Intron | XP_011544264.1 | |||
XM_017023732.1 | 4442 | Intron | XP_016879221.1 |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX1B - syntaxin 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052874.4 | 4442 | UTR 3 | NP_443106.1 | |||
XM_017022893.1 | 4442 | UTR 3 | XP_016878382.1 |
Set Membership: |
HapMap |