Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600846 | ||||||||||||||||||||
Literature Links: |
P2RX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
P2RX4 - purinergic receptor P2X 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256796.1 | 231 | UTR 5 | NP_001243725.1 | |||
NM_001261397.1 | 231 | UTR 5 | NP_001248326.1 | |||
NM_001261398.1 | 231 | UTR 5 | NP_001248327.1 | |||
NM_002560.2 | 231 | UTR 5 | NP_002551.2 | |||
XM_011538416.2 | 231 | Intron | XP_011536718.1 |