Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAAGGTCGGAGTTCTGACTGAA[A/G]TTCTTCCCACACTCATCACACTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611703 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF436 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF436 - zinc finger protein 436 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077195.1 | 1501 | Silent Mutation | AAC,AAT | N,N 314 | NP_001070663.1 | |
NM_030634.2 | 1501 | Silent Mutation | AAC,AAT | N,N 314 | NP_085137.1 | |
XM_011542201.1 | 1501 | Silent Mutation | AAC,AAT | N,N 296 | XP_011540503.1 |
ZNF436-AS1 - ZNF436 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |