Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602650 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SPOP - speckle type BTB/POZ protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007226.1 | 2354 | UTR 3 | NP_001007227.1 | |||
NM_001007227.1 | 2354 | UTR 3 | NP_001007228.1 | |||
NM_001007228.1 | 2354 | UTR 3 | NP_001007229.1 | |||
NM_001007229.1 | 2354 | UTR 3 | NP_001007230.1 | |||
NM_001007230.1 | 2354 | UTR 3 | NP_001007231.1 | |||
NM_003563.3 | 2354 | UTR 3 | NP_003554.1 | |||
XM_005257723.4 | 2354 | UTR 3 | XP_005257780.1 | |||
XM_005257724.4 | 2354 | UTR 3 | XP_005257781.1 | |||
XM_017025204.1 | 2354 | UTR 3 | XP_016880693.1 |
Set Membership: |
HapMap Validated |