Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602381 MIM: 616496 MIM: 601512 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NAB2 - NGFI-A binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005967.3 | 3570 | Intron | NP_005958.1 | |||
XM_005268894.2 | 3570 | Intron | XP_005268951.1 |
NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT6 - signal transducer and activator of transcription 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178078.1 | 3570 | UTR 3 | NP_001171549.1 | |||
NM_001178079.1 | 3570 | UTR 3 | NP_001171550.1 | |||
NM_001178080.1 | 3570 | UTR 3 | NP_001171551.1 | |||
NM_001178081.1 | 3570 | UTR 3 | NP_001171552.1 | |||
NM_003153.4 | 3570 | UTR 3 | NP_003144.3 | |||
XM_006719575.2 | 3570 | UTR 3 | XP_006719638.1 | |||
XM_011538703.2 | 3570 | UTR 3 | XP_011537005.1 | |||
XM_011538704.2 | 3570 | UTR 3 | XP_011537006.1 | |||
XM_011538705.2 | 3570 | UTR 3 | XP_011537007.1 | |||
XM_011538706.2 | 3570 | UTR 3 | XP_011537008.1 | |||
XM_011538707.2 | 3570 | UTR 3 | XP_011537009.1 | |||
XM_011538708.2 | 3570 | UTR 3 | XP_011537010.1 | |||
XM_011538709.2 | 3570 | UTR 3 | XP_011537011.1 |
Set Membership: |
HapMap Validated |