Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM206 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | Intron | NP_001185791.1 | ||||
NM_018252.2 | Intron | NP_060722.2 | ||||
XM_005273177.3 | Intron | XP_005273234.1 | ||||
XM_011509717.2 | Intron | XP_011508019.1 | ||||
XM_011509718.2 | Intron | XP_011508020.1 | ||||
XM_017001634.1 | Intron | XP_016857123.1 | ||||
XM_017001635.1 | Intron | XP_016857124.1 | ||||
XM_017001636.1 | Intron | XP_016857125.1 | ||||
XM_017001637.1 | Intron | XP_016857126.1 |
Set Membership: |
HapMap JSNP Validated |