Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608928 MIM: 605519 | ||||||||||||||||||||
Literature Links: |
EMILIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EMILIN2 - elastin microfibril interfacer 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LPIN2 - lipin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014646.2 | 6054 | UTR 3 | NP_055461.1 | |||
XM_005258177.4 | 6054 | UTR 3 | XP_005258234.1 | |||
XM_005258178.3 | 6054 | UTR 3 | XP_005258235.1 | |||
XM_005258179.4 | 6054 | UTR 3 | XP_005258236.1 | |||
XM_017026098.1 | 6054 | UTR 3 | XP_016881587.1 | |||
XM_017026099.1 | 6054 | UTR 3 | XP_016881588.1 |