Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCTCACATTCTGTAAATAATCA[A/G]TGCTAACCGTTTCATGTATGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613629 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927410 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101927410 - uncharacterized LOC101927410 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIEZO2 - piezo type mechanosensitive ion channel component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022068.3 | Intron | NP_071351.2 | ||||
XM_011525723.2 | Intron | XP_011524025.1 | ||||
XM_011525724.2 | Intron | XP_011524026.1 | ||||
XM_011525725.2 | Intron | XP_011524027.1 | ||||
XM_011525726.2 | Intron | XP_011524028.1 | ||||
XM_017025918.1 | Intron | XP_016881407.1 |