Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCCCATACAGCTATCAGCCCG[C/T]CCCCGATCCAGGGCATCCTTCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BISPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BISPR - BST2 interferon stimulated positive regulator (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BST2 - bone marrow stromal cell antigen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004335.3 | Intron | NP_004326.1 |
LOC105372343 - uncharacterized LOC105372343 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVB12A - multivesicular body subunit 12A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |