Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609694 MIM: 605110 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GMIP - GEM interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288998.1 | 3007 | UTR 3 | NP_001275927.1 | |||
NM_001288999.1 | 3007 | UTR 3 | NP_001275928.1 | |||
NM_016573.3 | 3007 | UTR 3 | NP_057657.2 | |||
XM_005259927.2 | 3007 | UTR 3 | XP_005259984.1 | |||
XM_011528048.2 | 3007 | UTR 3 | XP_011526350.1 | |||
XM_017026861.1 | 3007 | UTR 3 | XP_016882350.1 | |||
XM_017026862.1 | 3007 | Intron | XP_016882351.1 |
LPAR2 - lysophosphatidic acid receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004720.5 | 3007 | Intron | NP_004711.2 | |||
XM_011528421.2 | 3007 | Intron | XP_011526723.1 | |||
XM_017027468.1 | 3007 | Intron | XP_016882957.1 | |||
XM_017027469.1 | 3007 | Intron | XP_016882958.1 |
Set Membership: |
HapMap Validated |