Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREB1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GREB1L - growth regulation by estrogen in breast cancer 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142966.1 | Intron | NP_001136438.1 | ||||
XM_006722547.2 | Intron | XP_006722610.1 | ||||
XM_011526179.2 | Intron | XP_011524481.1 | ||||
XM_017025988.1 | Intron | XP_016881477.1 | ||||
XM_017025989.1 | Intron | XP_016881478.1 | ||||
XM_017025990.1 | Intron | XP_016881479.1 | ||||
XM_017025991.1 | Intron | XP_016881480.1 | ||||
XM_017025992.1 | Intron | XP_016881481.1 | ||||
XM_017025993.1 | Intron | XP_016881482.1 | ||||
XM_017025994.1 | Intron | XP_016881483.1 | ||||
XM_017025995.1 | Intron | XP_016881484.1 | ||||
XM_017025996.1 | Intron | XP_016881485.1 |
Set Membership: |
HapMap |