Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATACATGAGGATGAAGCCCAT[A/G]GCGCCCCGGTAGTATGCGGTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616133 MIM: 600128 MIM: 179490 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102725254 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102725254 - uncharacterized LOC102725254 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPV17L2 - MPV17 mitochondrial inner membrane protein like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE4C - phosphodiesterase 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3A - RAB3A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002866.4 | 451 | Silent Mutation | GCC,GCT | A,A 95 | NP_002857.1 | |
XM_011528164.1 | 451 | Silent Mutation | GCC,GCT | A,A 95 | XP_011526466.1 |