Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601270 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP4F3 - cytochrome P450 family 4 subfamily F member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000896.2 | Intron | NP_000887.2 | ||||
NM_001199208.1 | Intron | NP_001186137.1 | ||||
NM_001199209.1 | Intron | NP_001186138.1 | ||||
XM_005259911.4 | Intron | XP_005259968.1 | ||||
XM_011528014.2 | Intron | XP_011526316.1 | ||||
XM_017026814.1 | Intron | XP_016882303.1 | ||||
XM_017026815.1 | Intron | XP_016882304.1 |
Set Membership: |
HapMap |