Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAAATGCAAAAGAAGCCAGA[C/G]AGTAGACTTAACAACCATGAAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWF19L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CWF19L2 - CWF19-like 2, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152434.2 | 3095 | UTR 3 | NP_689647.2 | |||
XM_011542620.2 | 3095 | Intron | XP_011540922.1 | |||
XM_017017239.1 | 3095 | UTR 3 | XP_016872728.1 |
LOC105369477 - uncharacterized LOC105369477 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |