Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138246 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRIA4 - glutamate ionotropic receptor AMPA type subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000829.3 | Intron | NP_000820.3 | ||||
NM_001077243.2 | Intron | NP_001070711.2 | ||||
NM_001077244.1 | Intron | NP_001070712.1 | ||||
NM_001112812.1 | Intron | NP_001106283.1 | ||||
XM_005271518.3 | Intron | XP_005271575.1 | ||||
XM_006718823.1 | Intron | XP_006718886.1 | ||||
XM_011542775.2 | Intron | XP_011541077.1 | ||||
XM_011542776.2 | Intron | XP_011541078.1 | ||||
XM_011542777.2 | Intron | XP_011541079.1 | ||||
XM_017017609.1 | Intron | XP_016873098.1 | ||||
XM_017017610.1 | Intron | XP_016873099.1 | ||||
XM_017017611.1 | Intron | XP_016873100.1 |
Set Membership: |
HapMap |