Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNORD12 - small nucleolar RNA, C/D box 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD12B - small nucleolar RNA, C/D box 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD12C - small nucleolar RNA, C/D box 12C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFAS1 - ZNFX1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNFX1 - zinc finger NFX1-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021035.2 | Intron | NP_066363.1 | ||||
XM_006723836.1 | Intron | XP_006723899.1 | ||||
XM_011528924.1 | Intron | XP_011527226.1 | ||||
XM_017027971.1 | Intron | XP_016883460.1 |