Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613644 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF7IP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATF7IP - activating transcription factor 7 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286514.1 | 1761 | Intron | NP_001273443.1 | |||
NM_001286515.1 | 1761 | Intron | NP_001273444.1 | |||
NM_018179.4 | 1761 | Intron | NP_060649.3 | |||
NM_181352.1 | 1761 | Intron | NP_851997.1 | |||
XM_005253424.3 | 1761 | Intron | XP_005253481.1 | |||
XM_006719108.3 | 1761 | Intron | XP_006719171.1 | |||
XM_006719109.3 | 1761 | Intron | XP_006719172.1 | |||
XM_011520754.2 | 1761 | Intron | XP_011519056.1 | |||
XM_011520755.2 | 1761 | Intron | XP_011519057.1 | |||
XM_011520756.2 | 1761 | Intron | XP_011519058.1 | |||
XM_017019638.1 | 1761 | Intron | XP_016875127.1 | |||
XM_017019639.1 | 1761 | Intron | XP_016875128.1 |
PLBD1 - phospholipase B domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024829.5 | 1761 | Missense Mutation | CCA,GCA | P,A 534 | NP_079105.4 |
Set Membership: |
HapMap Validated |