Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609146 MIM: 604481 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6743 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6743 - microRNA 6743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIC8A - RIC8 guanine nucleotide exchange factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT3 - sirtuin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017524.2 | Intron | NP_001017524.1 | ||||
NM_012239.5 | Intron | NP_036371.1 | ||||
XM_005252835.1 | Intron | XP_005252892.1 | ||||
XM_011519956.1 | Intron | XP_011518258.1 | ||||
XM_011519957.1 | Intron | XP_011518259.1 | ||||
XM_017017428.1 | Intron | XP_016872917.1 | ||||
XM_017017429.1 | Intron | XP_016872918.1 | ||||
XM_017017430.1 | Intron | XP_016872919.1 | ||||
XM_017017431.1 | Intron | XP_016872920.1 |