Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCK1-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCK1-AS1 - NCK1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35G2 - solute carrier family 35 member G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097599.1 | Intron | NP_001091068.1 | ||||
NM_001097600.1 | Intron | NP_001091069.1 | ||||
NM_025246.2 | Intron | NP_079522.2 | ||||
XM_006713773.3 | Intron | XP_006713836.1 | ||||
XM_011513214.2 | Intron | XP_011511516.1 | ||||
XM_017007289.1 | Intron | XP_016862778.1 | ||||
XM_017007290.1 | Intron | XP_016862779.1 | ||||
XM_017007291.1 | Intron | XP_016862780.1 |