Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606241 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DICER1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DICER1 - dicer 1, ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195573.1 | Intron | NP_001182502.1 | ||||
NM_001271282.2 | Intron | NP_001258211.1 | ||||
NM_001291628.1 | Intron | NP_001278557.1 | ||||
NM_030621.4 | Intron | NP_085124.2 | ||||
NM_177438.2 | Intron | NP_803187.1 | ||||
XM_011536599.1 | Intron | XP_011534901.1 | ||||
XM_011536600.2 | Intron | XP_011534902.1 | ||||
XM_011536601.2 | Intron | XP_011534903.1 | ||||
XM_011536602.2 | Intron | XP_011534904.1 | ||||
XM_011536604.2 | Intron | XP_011534906.1 | ||||
XM_011536605.2 | Intron | XP_011534907.1 | ||||
XM_017021120.1 | Intron | XP_016876609.1 | ||||
XM_017021121.1 | Intron | XP_016876610.1 | ||||
XM_017021122.1 | Intron | XP_016876611.1 | ||||
XM_017021123.1 | Intron | XP_016876612.1 |