Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607663 MIM: 610693 MIM: 616283 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DDX25 - DEAD-box helicase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013264.4 | Intron | NP_037396.3 | ||||
XM_011542790.2 | Intron | XP_011541092.1 | ||||
XM_011542791.2 | Intron | XP_011541093.1 | ||||
XM_017017626.1 | Intron | XP_016873115.1 |
HYLS1 - HYLS1, centriolar and ciliogenesis associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUS3 - pseudouridylate synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271985.1 | Intron | NP_001258914.1 | ||||
NM_031307.3 | Intron | NP_112597.3 | ||||
XM_005271687.3 | Intron | XP_005271744.1 | ||||
XM_005271688.3 | Intron | XP_005271745.1 |
Set Membership: |
HapMap Validated |