Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605621 MIM: 600508 | ||||||||||||||||||||
Literature Links: |
IL20RB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL20RB - interleukin 20 receptor subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144717.3 | 645 | Intron | NP_653318.2 | |||
XM_006713665.3 | 645 | UTR 5 | XP_006713728.1 | |||
XM_011512910.2 | 645 | Intron | XP_011511212.1 | |||
XM_011512911.2 | 645 | Intron | XP_011511213.1 | |||
XM_017006632.1 | 645 | Intron | XP_016862121.1 |
NCK1 - NCK adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |