Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATCTTTTCTACCTGTGGCTCTC[C/T]ACCAATTGTTACTCTCTTGTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC103611081 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC103611081 - uncharacterized LOC103611081 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX19 - sorting nexin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301089.1 | 3684 | UTR 3 | NP_001288018.1 | |||
NM_014758.2 | 3684 | UTR 3 | NP_055573.2 | |||
XM_005271546.4 | 3684 | UTR 3 | XP_005271603.1 | |||
XM_011542819.2 | 3684 | UTR 3 | XP_011541121.1 | |||
XM_011542820.2 | 3684 | UTR 3 | XP_011541122.1 | |||
XM_011542821.2 | 3684 | UTR 3 | XP_011541123.1 | |||
XM_011542822.2 | 3684 | Intron | XP_011541124.1 | |||
XM_011542824.2 | 3684 | UTR 3 | XP_011541126.1 | |||
XM_011542825.2 | 3684 | UTR 3 | XP_011541127.1 | |||
XM_011542826.2 | 3684 | UTR 3 | XP_011541128.1 | |||
XM_011542827.2 | 3684 | UTR 3 | XP_011541129.1 | |||
XM_017017725.1 | 3684 | UTR 3 | XP_016873214.1 | |||
XM_017017726.1 | 3684 | UTR 3 | XP_016873215.1 |
Set Membership: |
HapMap |