Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142940 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HMGCS1 - 3-hydroxy-3-methylglutaryl-CoA synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098272.2 | 2216 | UTR 3 | NP_001091742.1 | |||
NM_001324219.1 | 2216 | UTR 3 | NP_001311148.1 | |||
NM_001324220.1 | 2216 | UTR 3 | NP_001311149.1 | |||
NM_001324222.1 | 2216 | UTR 3 | NP_001311151.1 | |||
NM_001324223.1 | 2216 | UTR 3 | NP_001311152.1 | |||
NM_001324224.1 | 2216 | UTR 3 | NP_001311153.1 | |||
NM_002130.7 | 2216 | UTR 3 | NP_002121.4 | |||
XM_011514036.2 | 2216 | UTR 3 | XP_011512338.1 | |||
XM_017009412.1 | 2216 | UTR 3 | XP_016864901.1 |
NIM1K - NIM1 serine/threonine protein kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |