Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCAGGAGTTTGACACCAGCTT[G/T]GGCAATATAGGGAGACCCCGTCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601045 | ||||||||||||||||||||
Literature Links: |
CTNND1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTNND1 - catenin delta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085458.1 | Intron | NP_001078927.1 | ||||
NM_001085459.1 | Intron | NP_001078928.1 | ||||
NM_001085460.1 | Intron | NP_001078929.1 | ||||
NM_001085461.1 | Intron | NP_001078930.1 | ||||
NM_001085462.1 | Intron | NP_001078931.1 | ||||
NM_001085463.1 | Intron | NP_001078932.1 | ||||
NM_001085464.1 | Intron | NP_001078933.1 | ||||
NM_001085465.1 | Intron | NP_001078934.1 | ||||
NM_001085466.1 | Intron | NP_001078935.1 | ||||
NM_001085467.1 | Intron | NP_001078936.1 | ||||
NM_001085468.1 | Intron | NP_001078937.1 | ||||
NM_001085469.1 | Intron | NP_001078938.1 | ||||
NM_001206883.1 | Intron | NP_001193812.1 | ||||
NM_001206884.1 | Intron | NP_001193813.1 | ||||
NM_001206885.1 | Intron | NP_001193814.1 | ||||
NM_001206886.1 | Intron | NP_001193815.1 | ||||
NM_001206887.1 | Intron | NP_001193816.1 | ||||
NM_001206888.1 | Intron | NP_001193817.1 | ||||
NM_001206889.1 | Intron | NP_001193818.1 | ||||
NM_001206890.1 | Intron | NP_001193819.1 | ||||
NM_001206891.1 | Intron | NP_001193820.1 | ||||
NM_001331.2 | Intron | NP_001322.1 |
TMX2-CTNND1 - TMX2-CTNND1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |