Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGAGAGCTCGGAAGAGGAGGAA[G/T]AAGCAGAAGCCGAAACCGCCAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606847 | ||||||||||||||||||||
Literature Links: |
TCOF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCOF1 - treacle ribosome biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000356.3 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_000347.2 | |
NM_001008657.2 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_001008657.1 | |
NM_001135243.1 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_001128715.1 | |
NM_001135244.1 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_001128716.1 | |
NM_001135245.1 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_001128717.1 | |
NM_001195141.1 | 385 | Nonsense Mutation | GAA,TAA | E,* 93 | NP_001182070.1 |