Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142790 MIM: 606847 | ||||||||||||||||||||
Literature Links: |
CD74 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD74 - CD74 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025158.2 | 1381 | UTR 3 | NP_001020329.1 | |||
NM_001025159.2 | 1381 | UTR 3 | NP_001020330.1 | |||
NM_004355.3 | 1381 | UTR 3 | NP_004346.1 | |||
XM_017010089.1 | 1381 | UTR 3 | XP_016865578.1 | |||
XM_017010090.1 | 1381 | UTR 3 | XP_016865579.1 |
TCOF1 - treacle ribosome biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |