Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGTTTCTACAAAAACCTAGGCA[G/T]GTTAAAAAGACGTGCTTCATTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611197 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAKMIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
JAKMIP2 - janus kinase and microtubule interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270934.1 | 6968 | UTR 3 | NP_001257863.1 | |||
NM_001270941.1 | 6968 | UTR 3 | NP_001257870.1 | |||
NM_001282282.1 | 6968 | UTR 3 | NP_001269211.1 | |||
NM_014790.4 | 6968 | UTR 3 | NP_055605.2 | |||
XM_017010097.1 | 6968 | UTR 3 | XP_016865586.1 | |||
XM_017010098.1 | 6968 | UTR 3 | XP_016865587.1 |
JAKMIP2-AS1 - JAKMIP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |