Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608150 MIM: 608500 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPHLN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPHLN1 - periphilin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRICKLE1 - prickle planar cell polarity protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144881.1 | 3884 | UTR 3 | NP_001138353.1 | |||
NM_001144882.1 | 3884 | UTR 3 | NP_001138354.1 | |||
NM_001144883.1 | 3884 | UTR 3 | NP_001138355.1 | |||
NM_153026.2 | 3884 | UTR 3 | NP_694571.2 | |||
XM_011537946.1 | 3884 | UTR 3 | XP_011536248.1 | |||
XM_011537947.2 | 3884 | UTR 3 | XP_011536249.1 | |||
XM_017018838.1 | 3884 | UTR 3 | XP_016874327.1 | |||
XM_017018839.1 | 3884 | UTR 3 | XP_016874328.1 | |||
XM_017018840.1 | 3884 | UTR 3 | XP_016874329.1 |
Set Membership: |
HapMap |