Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120105 | ||||||||||||||||||||
Literature Links: |
CLPS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLPS - colipase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252597.1 | 435 | Missense Mutation | GGC,GTC | G,V 80 | NP_001239526.1 | |
NM_001252598.1 | 435 | Missense Mutation | GGC,GTC | G,V 53 | NP_001239527.1 | |
NM_001832.3 | 435 | Missense Mutation | GGC,GTC | G,V 94 | NP_001823.1 |
CLPSL1 - colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |