Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608508 MIM: 604628 MIM: 603236 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYBA - cytochrome b-245 alpha chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000101.3 | 592 | Missense Mutation | GCG,GTG | A,V 174 | NP_000092.2 | |
XM_011522905.2 | 592 | Intron | XP_011521207.1 |
IL17C - interleukin 17C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVD - mevalonate diphosphate decarboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |