Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142991 MIM: 142988 MIM: 142989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVX2 - even-skipped homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD12 - homeobox D12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD13 - homeobox D13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000523.3 | 2447 | Intron | NP_000514.2 | |||
XM_011511068.2 | 2447 | Intron | XP_011509370.1 | |||
XM_011511069.2 | 2447 | UTR 3 | XP_011509371.1 |