Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAGGAAAAGGTGGATATAAAG[G/T]GGAACCTGTGGGAAAGAGGCAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACIN1 - apoptotic chromatin condensation inducer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164814.1 | 4634 | UTR 3 | NP_001158286.1 | |||
NM_001164815.1 | 4634 | UTR 3 | NP_001158287.1 | |||
NM_001164816.1 | 4634 | UTR 3 | NP_001158288.1 | |||
NM_001164817.1 | 4634 | UTR 3 | NP_001158289.1 | |||
NM_014977.3 | 4634 | UTR 3 | NP_055792.1 | |||
XM_005267415.3 | 4634 | UTR 3 | XP_005267472.1 | |||
XM_005267416.3 | 4634 | UTR 3 | XP_005267473.1 | |||
XM_005267418.1 | 4634 | UTR 3 | XP_005267475.1 | |||
XM_006720081.3 | 4634 | Intron | XP_006720144.1 | |||
XM_011536570.2 | 4634 | Intron | XP_011534872.1 |
CDH24 - cadherin 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105370705 - collagen alpha-2(I) chain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011537450.1 | 4634 | Intron | XP_011535752.1 |