Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607376 | ||||||||||||||||||||
Literature Links: |
DCTN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCTN2 - dynactin subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261412.1 | 854 | Missense Mutation | ACT,CCT | T,P 198 | NP_001248341.1 | |
NM_001261413.1 | 854 | Missense Mutation | ACT,CCT | T,P 196 | NP_001248342.1 | |
NM_006400.4 | 854 | Missense Mutation | ACT,CCT | T,P 201 | NP_006391.1 | |
XM_011537761.2 | 854 | Missense Mutation | ACT,CCT | T,P 109 | XP_011536063.1 |
MBD6 - methyl-CpG binding domain protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |