Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607376 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCTN2 - dynactin subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261412.1 | 1515 | UTR 3 | NP_001248341.1 | |||
NM_001261413.1 | 1515 | UTR 3 | NP_001248342.1 | |||
NM_006400.4 | 1515 | UTR 3 | NP_006391.1 | |||
XM_011537761.2 | 1515 | UTR 3 | XP_011536063.1 |
MBD6 - methyl-CpG binding domain protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052897.3 | 1515 | Intron | NP_443129.3 | |||
XM_005268617.1 | 1515 | Intron | XP_005268674.1 | |||
XM_005268618.1 | 1515 | Intron | XP_005268675.1 | |||
XM_005268620.2 | 1515 | Intron | XP_005268677.1 | |||
XM_006719217.1 | 1515 | Intron | XP_006719280.1 | |||
XM_006719218.1 | 1515 | Intron | XP_006719281.1 | |||
XM_011537851.2 | 1515 | Intron | XP_011536153.1 |
Set Membership: |
HapMap |