Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606099 | ||||||||||||||||||||
Literature Links: |
HEATR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEATR1 - HEAT repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018072.5 | 5195 | Silent Mutation | CCC,CCT | P,P 2108 | NP_060542.4 |
LGALS8 - galectin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006499.4 | 5195 | UTR 3 | NP_006490.3 | |||
NM_201543.2 | 5195 | UTR 3 | NP_963837.1 | |||
NM_201544.2 | 5195 | UTR 3 | NP_963838.1 | |||
NM_201545.2 | 5195 | UTR 3 | NP_963839.1 | |||
XM_011544188.2 | 5195 | Intron | XP_011542490.1 | |||
XM_017001274.1 | 5195 | Intron | XP_016856763.1 | |||
XM_017001275.1 | 5195 | Intron | XP_016856764.1 |