Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGCCGGGCTGCCTGCCCCACTT[G/T]AAGGCAGGCCTCCCCAGAGGCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102480 MIM: 606230 | ||||||||||||||||||||
Literature Links: |
ACR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACR - acrosin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097.2 | 510 | Missense Mutation | TTG,TTT | L,F 166 | NP_001088.2 |
LOC105373100 - uncharacterized LOC105373100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHANK3 - SH3 and multiple ankyrin repeat domains 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |