Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186790 MIM: 186740 | ||||||||||||||||||||
Literature Links: |
CD3D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD3D - CD3d molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000732.4 | 517 | Missense Mutation | GCA,GGA | A,G 127 | NP_000723.1 | |
NM_001040651.1 | 517 | Intron | NP_001035741.1 | |||
XM_017018543.1 | 517 | Missense Mutation | GCA,GGA | A,G 127 | XP_016874032.1 |
CD3G - CD3g molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |