Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600594 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGCR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DGCR11 - DiGeorge syndrome critical region gene 11 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGCR2 - DiGeorge syndrome critical region gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173533.1 | 2131 | UTR 3 | NP_001167004.1 | |||
NM_001173534.1 | 2131 | UTR 3 | NP_001167005.1 | |||
NM_001184781.1 | 2131 | UTR 3 | NP_001171710.1 | |||
NM_005137.2 | 2131 | UTR 3 | NP_005128.1 |
DGCR5 - DiGeorge syndrome critical region gene 5 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |