Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601405 MIM: 616450 | ||||||||||||||||||||
Literature Links: |
CTRC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTRC - chymotrypsin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007272.2 | 36 | Missense Mutation | ACT,TCT | T,S 5 | NP_009203.2 | |
XM_011540550.1 | 36 | Missense Mutation | ACT,TCT | T,S 5 | XP_011538852.1 |
EFHD2 - EF-hand domain family member D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |