Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609030 | ||||||||||||||||||||
Literature Links: |
DGCR8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGCR8 - DGCR8 microprocessor complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190326.1 | Intron | NP_001177255.1 | ||||
NM_022720.6 | Intron | NP_073557.3 | ||||
XM_006724268.2 | Intron | XP_006724331.1 |
MIR1306 - microRNA 1306 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3618 - microRNA 3618 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |