Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103180 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARF1 - ADP ribosylation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024226.1 | Intron | NP_001019397.1 | ||||
NM_001024227.1 | Intron | NP_001019398.1 | ||||
NM_001024228.1 | Intron | NP_001019399.1 | ||||
NM_001658.3 | Intron | NP_001649.1 | ||||
XM_017001233.1 | Intron | XP_016856722.1 |
C1orf35 - chromosome 1 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3620 - microRNA 3620 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |