Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600237 MIM: 605089 MIM: 601754 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C22orf39 - chromosome 22 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166242.1 | 1443 | UTR 3 | NP_001159714.1 | |||
NM_173793.4 | 1443 | Intron | NP_776154.3 |
HIRA - histone cell cycle regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL40 - mitochondrial ribosomal protein L40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |