Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC157 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC157 - coiled-coil domain containing 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017437.3 | 1746 | Intron | NP_001017437.2 | |||
NM_001318334.1 | 1746 | Intron | NP_001305263.1 | |||
NM_001318335.1 | 1746 | Intron | NP_001305264.1 | |||
XM_006724278.3 | 1746 | Intron | XP_006724341.1 | |||
XM_006724279.3 | 1746 | Intron | XP_006724342.1 | |||
XM_006724280.3 | 1746 | Intron | XP_006724343.1 | |||
XM_011530263.2 | 1746 | Intron | XP_011528565.1 | |||
XM_011530266.2 | 1746 | Intron | XP_011528568.1 | |||
XM_011530267.2 | 1746 | Intron | XP_011528569.1 | |||
XM_011530268.2 | 1746 | Intron | XP_011528570.1 | |||
XM_011530269.2 | 1746 | Intron | XP_011528571.1 | |||
XM_011530270.2 | 1746 | Intron | XP_011528572.1 |
KIAA1656 - KIAA1656 protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF215 - ring finger protein 215 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017981.1 | 1746 | UTR 3 | NP_001017981.1 |