Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTAGCCATCCCCAATAAACCCA[C/T]GAAGTTCAGCCTGAACAGCATAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603126 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507599 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507599 - uncharacterized LOC100507599 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPST2 - tyrosylprotein sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008566.1 | 3457 | Intron | NP_001008566.1 | |||
NM_003595.3 | 3457 | Intron | NP_003586.3 | |||
XM_006724338.3 | 3457 | UTR 3 | XP_006724401.1 |
Set Membership: |
HapMap |